Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024336.1 | NP_001019507.1 | 2 | 128 | ||
GenBank mRNA | BC079242.1 | - | 2 | 128 | 7132665 |
Gene Symbol: Tuba3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040008.1 | NP_001035097.1 | 2 | 124 | ||
GenBank mRNA | BC079395.1 | - | 2 | 124 | 7112099 |