Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004798.3 | NP_004789.1 | 7 | 2088 | ||
GenBank mRNA | AB002357.1 | - | 7 | 2075 | ||
AK098431.1 | - | 6 | 633 | |||
AK299877.1 | - | 7 | 1011 | |||
BC136310.1 | - | 6 | 1981 | 9020297 | ||
BC136311.1 | - | 6 | 1981 | 9020298 |