Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171802.1 | NP_001165273.1 | 3, 4 | 352 | ||
XM_006237906.2 | XP_006237968.1 | 3, 4 | 363 | |||
GenBank mRNA | BC089993.1 | - | 3, 4 | 322 | 7319652 | |
CB805415.1 | - | 3, 4 | 352 |