Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271238.1 | NP_001258167.1 | 9 | 986 | ||
XM_008763596.1 | XP_008761818.1 | 7 | 1230 |