Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031689.2 | NP_001026859.1 | 8 | 1523 | ||
NM_001321546.1 | NP_001308475.1 | 8 | 1526 | |||
XM_011518072.2 | XP_011516374.1 | 8 | 1512 | |||
XM_017015319.1 | XP_016870808.1 | 8 | 1512 | |||
GenBank mRNA | AF083395.1 | - | 7 | 927 | ||
AF145020.1 | - | 8 | 1322 | |||
AJ238243.1 | - | 7 | 955 | |||
AK001642.1 | - | 8 | 1226 | |||
AK002143.1 | - | 8 | 1286 | |||
AK022761.1 | - | 8 | 1201 | |||
AK223544.1 | - | 7 | 958 | |||
AL133608.1 | - | 8 | 1516 | |||
BC032551.1 | - | 8 | 1205 | 5555038 |