Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004800.2 | NP_004791.1 | 6 | 936 | ||
GenBank mRNA | AK093507.1 | - | 6 | 533 | ||
AK290514.1 | - | 6 | 891 | |||
AK315210.1 | - | 6 | 891 | |||
BC110656.1 | - | 6 | 893 | 3855486 | ||
BC130293.1 | - | 6 | 770 | 40146324 | ||
U81006.1 | - | 6 | 836 |