Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034949.1 | NP_001030121.1 | 8, 9 | 982 | ||
XM_006239262.1 | XP_006239324.1 | 8, 9 | 2407 | |||
GenBank mRNA | BC099755.1 | - | 8, 9 | 982 | 5623720 |