Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025773.1 | NP_001020944.1 | 5, 6 | 849 | ||
XM_006239471.2 | XP_006239533.1 | 6, 7 | 1269 | |||
XM_006239472.2 | XP_006239534.1 | 5, 6 | 1702 | |||
XM_008764282.1 | XP_008762504.1 | 5, 6 | 1039 | |||
XM_008764283.1 | XP_008762505.1 | 6, 7 | 1290 | |||
GenBank mRNA | AB164643.1 | - | 4, 5 | 425 | ||
BC097483.1 | - | 5, 6 | 849 | 7459228 |