Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024349.2 | NP_001019520.2 | 2, 3 | 327 | ||
XM_008764373.1 | XP_008762595.1 | 2, 3 | 1582 | |||
XM_008764374.1 | XP_008762596.1 | 3, 4 | 1500 | |||
GenBank mRNA | BC088265.1 | - | 2, 3 | 328 | 7377562 |