Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012203.1 | NP_036335.1 | 8 | 853 | ||
XM_005251631.1 | XP_005251688.1 | 5 | 546 | |||
XM_011518073.2 | XP_011516375.1 | 8 | 1227 | |||
XM_017015320.1 | XP_016870809.1 | 8 | 897 | |||
XM_017015321.1 | XP_016870810.1 | 8 | 897 | |||
XM_017015322.1 | XP_016870811.1 | 5 | 546 | |||
XM_017015323.1 | XP_016870812.1 | 8 | 1227 | |||
GenBank mRNA | AF113215.1 | - | 8 | 861 | ||
AF113251.1 | - | 8 | 871 | |||
AF134895.1 | - | 8 | 853 | |||
AF146018.1 | - | 8 | 852 | |||
AK024386.1 | - | 1 | 4379 | |||
AK025978.1 | - | 8 | 897 | |||
AK026752.1 | - | 3 | 1514 | |||
AK315690.1 | - | 8 | 867 | |||
BC000605.2 | - | 8 | 832 | 3347232 | ||
BC003131.1 | - | 8 | 853 | |||
BC088360.1 | - | 8 | 851 |