Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271107.1 | NP_001258036.1 | 1 | 731 | ||
GenBank mRNA | BC098054.1 | - | 1 | 123 |