Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729635.3 | XP_002729681.1 | 4 | 934 | ||
XM_576044.5 | XP_576044.1 | 4 | 1045 |