Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234425.2 | XP_006234487.1 | 17 | 1965 | ||
XM_008761965.1 | XP_008760187.1 | 17 | 1980 | |||
XM_008761966.1 | XP_008760188.1 | 17 | 1967 | |||
XM_008761967.1 | XP_008760189.1 | 17 | 1977 | |||
XM_342453.7 | XP_342454.3 | 17 | 1967 | |||
GenBank mRNA | FQ231274.1 | - | 17 | 1894 | ||
FQ231579.1 | - | 17 | 1894 |
Gene Symbol: LOC500684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047959.1 | NP_001041424.1 | 2 | 1712 | ||
GenBank mRNA | AY325221.1 | - | 2 | 1712 |