Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100872.1 | NP_001094342.1 | 8 | 1745 | ||
XM_006240303.2 | XP_006240365.1 | 8 | 1968 |