Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1566220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240360.2 | XP_006240422.1 | 12 | 2561 | ||
XM_006240361.2 | XP_006240423.1 | 13 | 2618 | |||
XM_006240362.2 | XP_006240424.1 | 13 | 2627 | |||
XM_006240363.2 | XP_006240425.2 | 13 | 2707 |