Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004256.3 | NP_004247.2 | 5 | 921 | ||
XM_017007519.1 | XP_016863008.1 | 4 | 667 | |||
GenBank mRNA | AB010438.1 | - | 5 | 888 | ||
AK298375.1 | - | 4 | 667 | |||
AK315301.1 | - | 5 | 916 | |||
BC035973.1 | - | 5 | 916 | 4609915 |