Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053971.3 | NP_446423.2 | 2, 3 | 374 | ||
XM_006249377.1 | XP_006249439.1 | 3, 4 | 393 | |||
XM_006249378.2 | XP_006249440.1 | 3, 4 | 427 | |||
GenBank mRNA | BC061784.1 | - | 2, 3 | 370 | ||
BC078761.1 | - | 2, 3 | 377 | 7123086 | ||
BC097295.1 | - | 2, 3 | 378 | 7446680 | ||
FQ209419.1 | - | 1, 2 | 52 | |||
FQ213055.1 | - | 1, 2 | 52 | |||
FQ213078.1 | - | 1, 2 | 52 | |||
FQ216758.1 | - | 1, 2 | 52 | |||
FQ217744.1 | - | 1, 2 | 52 | |||
FQ219956.1 | - | 1, 2 | 52 | |||
FQ219982.1 | - | 1, 2 | 52 | |||
FQ220180.1 | - | 1, 2 | 52 | |||
FQ220346.1 | - | 1, 2 | 52 | |||
FQ220362.1 | - | 1, 2 | 52 | |||
FQ220471.1 | - | 1, 2 | 52 | |||
FQ220678.1 | - | 1, 2 | 52 | |||
FQ220699.1 | - | 1, 2 | 52 | |||
FQ220887.1 | - | 1, 2 | 52 | |||
FQ220959.1 | - | 1, 2 | 52 | |||
FQ221615.1 | - | 1, 2 | 52 | |||
FQ222080.1 | - | 1, 2 | 52 | |||
FQ222331.1 | - | 1, 2 | 52 | |||
FQ222465.1 | - | 1, 2 | 52 | |||
FQ222658.1 | - | 1, 2 | 52 | |||
FQ223301.1 | - | 1, 2 | 52 | |||
FQ226632.1 | - | 1, 2 | 52 | |||
FQ227060.1 | - | 1, 2 | 52 | |||
FQ227100.1 | - | 1, 2 | 52 | |||
FQ227955.1 | - | 1, 2 | 52 | |||
FQ227974.1 | - | 1, 2 | 52 | |||
FQ228583.1 | - | 1, 2 | 52 | |||
FQ228665.1 | - | 1, 2 | 52 | |||
FQ229345.1 | - | 1, 2 | 52 | |||
FQ229355.1 | - | 1, 2 | 52 | |||
FQ229484.1 | - | 3, 4 | 392 | |||
FQ229571.1 | - | 1, 2 | 52 | |||
FQ229624.1 | - | 1, 2 | 52 | |||
FQ229675.1 | - | 1, 2 | 52 | |||
FQ229700.1 | - | 1, 2 | 52 | |||
FQ229820.1 | - | 1, 2 | 52 | |||
FQ229854.1 | - | 1, 2 | 52 | |||
FQ229857.1 | - | 1, 2 | 52 | |||
FQ229943.1 | - | 3, 4 | 391 | |||
FQ230171.1 | - | 1, 2 | 52 | |||
FQ232778.1 | - | 1, 2 | 52 | |||
FQ233198.1 | - | 1, 2 | 52 | |||
FQ233475.1 | - | 1, 2 | 52 | |||
FQ234880.1 | - | 3, 4 | 391 | |||
FQ235095.1 | - | 1, 2 | 52 | |||
X87107.1 | - | 3, 4 | 388 |