Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053971.3 | NP_446423.2 | 1 | 123 | ||
XM_006249377.1 | XP_006249439.1 | 2 | 142 | |||
XM_006249378.2 | XP_006249440.1 | 2 | 176 | |||
GenBank mRNA | BC061784.1 | - | 1 | 119 | ||
BC078761.1 | - | 1 | 126 | 7123086 | ||
BC097295.1 | - | 1 | 127 | 7446680 | ||
FQ229484.1 | - | 2 | 141 | |||
FQ229943.1 | - | 2 | 140 | |||
FQ234880.1 | - | 2 | 140 | |||
X87107.1 | - | 2 | 137 |