Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109672.1 | NP_001103142.1 | 9 | 882 | ||
XM_006240549.2 | XP_006240611.1 | 10 | 1125 | |||
XM_006240551.2 | XP_006240613.1 | 8 | 1127 | |||
GenBank mRNA | BC129121.1 | - | 9 | 882 | 7125896 |