Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109285.1 | NP_001102755.1 | 2 | 1511 | ||
XM_006240985.2 | XP_006241047.1 | 2 | 1697 | |||
XM_008765178.1 | XP_008763400.1 | 2 | 1433 |