Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130575.1 | NP_001124047.1 | 1, 2 | 1102 | ||
XM_008765455.1 | XP_008763677.1 | 1, 2 | 551 |