Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044270.2 | NP_001037735.1 | 4 | 1528 | ||
GenBank mRNA | BC060572.1 | - | 4 | 1532 | 6920989 | |
BC076379.1 | - | 4 | 1514 | 7106502 | ||
FQ212426.1 | - | 4 | 1523 | |||
FQ212740.1 | - | 4 | 1523 | |||
FQ213098.1 | - | 4 | 1528 | |||
FQ213226.1 | - | 4 | 1526 | |||
FQ213439.1 | - | 4 | 1525 | |||
FQ213947.1 | - | 4 | 1524 | |||
FQ214075.1 | - | 4 | 1526 | |||
FQ214100.1 | - | 4 | 1523 | |||
FQ214301.1 | - | 4 | 1527 | |||
FQ223081.1 | - | 4 | 1528 | |||
FQ225046.1 | - | 4 | 1532 | |||
FQ225339.1 | - | 4 | 1522 | |||
FQ225425.1 | - | 4 | 1528 | |||
FQ231550.1 | - | 4 | 1524 | |||
FQ232160.1 | - | 3 | 1433 | |||
FQ232527.1 | - | 4 | 1528 | |||
FQ233000.1 | - | 4 | 1525 | |||
FQ233566.1 | - | 4 | 1526 | |||
FQ234135.1 | - | 4 | 1523 | |||
V01226.1 | - | 1 | 511 |