Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109288.1 | NP_001102758.1 | 4, 5 | 442 | ||
XM_008765777.1 | XP_008763999.1 | 4, 5 | 404 |