Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134634.2 | NP_001128106.2 | 10 | 2329 | ||
XM_006242578.2 | XP_006242640.1 | 9 | 2321 | |||
XM_008765969.1 | XP_008764191.1 | 8 | 2135 | |||
XM_008765970.1 | XP_008764192.1 | 9 | 2196 | |||
XM_008765971.1 | XP_008764193.1 | 8 | 2139 | |||
XM_008765972.1 | XP_008764194.1 | 9 | 2200 | |||
XM_008765973.1 | XP_008764195.1 | 7 | 2077 | |||
XM_008765974.1 | XP_008764196.1 | 7 | 2081 | |||
XM_008765975.1 | XP_008764197.1 | 8 | 2111 | |||
XM_008765976.1 | XP_008764198.1 | 7 | 1973 | |||
XM_008765977.1 | XP_008764199.1 | 7 | 1949 | |||
GenBank mRNA | FQ211838.1 | - | 8 | 2115 |