Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134634.2 | NP_001128106.2 | 10 | 1115 | ||
XM_006242578.2 | XP_006242640.1 | 9 | 1107 | |||
XM_008765969.1 | XP_008764191.1 | 8 | 921 | |||
XM_008765970.1 | XP_008764192.1 | 9 | 982 | |||
XM_008765971.1 | XP_008764193.1 | 8 | 925 | |||
XM_008765972.1 | XP_008764194.1 | 9 | 986 | |||
XM_008765973.1 | XP_008764195.1 | 7 | 863 | |||
XM_008765974.1 | XP_008764196.1 | 7 | 867 | |||
XM_008765975.1 | XP_008764197.1 | 8 | 897 | |||
XM_008765976.1 | XP_008764198.1 | 7 | 759 | |||
XM_008765977.1 | XP_008764199.1 | 7 | 735 | |||
GenBank mRNA | BU671127.1 | - | 6 | 485 | ||
FQ211838.1 | - | 8 | 902 |