Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004260.3 | NP_004251.3 | 15 | 2762 | ||
XM_011517384.2 | XP_011515686.1 | 15 | 2564 | |||
XM_017013991.1 | XP_016869480.1 | 14 | 2857 | |||
XM_017013992.1 | XP_016869481.1 | 14 | 2856 | |||
XM_017013993.1 | XP_016869482.1 | 15 | 2766 | |||
XM_017013994.1 | XP_016869483.1 | 15 | 2762 | |||
XM_017013995.1 | XP_016869484.1 | 15 | 2764 | |||
XM_017013996.1 | XP_016869485.1 | 14 | 2855 | |||
XM_017013997.1 | XP_016869486.1 | 14 | 2659 | |||
XM_017013998.1 | XP_016869487.1 | 14 | 2854 | |||
XM_017013999.1 | XP_016869488.1 | 15 | 2568 | |||
XM_017014000.1 | XP_016869489.1 | 10 | 1768 | |||
XM_017014001.1 | XP_016869490.1 | 13 | 2858 | |||
GenBank mRNA | AB006532.1 | - | 15 | 2720 | ||
BC011602.2 | - | 8 | 1321 | |||
BC013277.2 | - | 6 | 1059 | 3634266 | ||
KJ147518.1 | - | 11 | 2069 |