Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC500959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033072.1 | NP_001028244.1 | 1 | 311 | ||
GenBank mRNA | AY461585.1 | - | 1 | 311 |