Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024359.1 | NP_001019530.1 | 4 | 765 | ||
GenBank mRNA | AY634367.1 | - | 4 | 765 |