Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109292.1 | NP_001102762.1 | 5 | 743 | ||
XM_006242950.2 | XP_006243012.1 | 5 | 743 | |||
XM_006242951.2 | XP_006243013.1 | 5 | 757 | |||
XM_006242952.2 | XP_006243014.1 | 5 | 752 | |||
XM_006242954.2 | XP_006243016.1 | 5 | 1026 |