Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143995.2 | NP_001137467.1 | 9 | 1257 | ||
NM_001307951.1 | NP_001294880.1 | 8 | 1243 | |||
NM_004811.2 | NP_004802.1 | 9 | 1194 | |||
XM_006718750.1 | XP_006718813.1 | 9 | 1290 | |||
XM_011545388.2 | XP_011543690.1 | 8 | 1914 | |||
XM_011545390.2 | XP_011543692.1 | 8 | 2224 | |||
XM_011545391.2 | XP_011543693.1 | 8 | 1190 | |||
GenBank mRNA | AF062075.1 | - | 9 | 1142 | ||
AK223165.1 | - | 9 | 1133 | |||
AK299264.1 | - | 8 | 1224 | |||
AK300955.1 | - | 9 | 1184 | |||
AK313306.1 | - | 9 | 1127 | |||
BC019035.2 | - | 9 | 1116 | 4575069 | ||
CR536531.1 | - | 8 | 1049 |