Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005455.4 | NP_005446.2 | 3 | 487 | ||
NM_203350.2 | NP_976225.1 | 3 | 487 | |||
GenBank mRNA | AB209568.1 | - | 3 | 200 | ||
AF065391.1 | - | 3 | 227 | |||
AF065392.1 | - | 3 | 227 | |||
AK222858.1 | - | 3 | 212 | |||
AL136945.1 | - | 3 | 209 | |||
AU100296.1 | - | 3 | 261 | |||
BC033648.1 | - | 3 | 233 | |||
BC039814.1 | - | 3 | 233 | 6062647 | ||
DA523601.1 | - | 3 | 487 |