Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il20rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173438.1 | NP_001166909.1 | 3 | 377 | ||
GenBank mRNA | DQ222846.1 | - | 3 | 377 |