Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277434.1 | NP_001264363.1 | 6 | 1140 | ||
GenBank mRNA | FQ227817.1 | - | 4 | 424 | ||
FQ228265.1 | - | 7 | 764 | |||
FQ230804.1 | - | 4 | 424 |