Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244725.2 | XP_006244787.1 | 3 | 1819 | ||
XM_576539.5 | XP_576539.3 | 2 | 1718 |