Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037767.2 | NP_001032856.2 | 1 | 115 | ||
GenBank mRNA | BC127445.1 | - | 1 | 44 | ||
FM029541.1 | - | 1 | 115 |