Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750700.3 | XP_003750748.1 | 2 | 305 | ||
XM_006244941.2 | XP_006245003.1 | 2 | 306 | |||
XM_008767154.1 | XP_008765376.1 | 2 | 307 | |||
XM_008767155.1 | XP_008765377.1 | 2 | 306 |
Gene Symbol: Boll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113370.1 | NP_001106841.1 | 2 | 296 | ||
XM_006244929.2 | XP_006244991.1 | 2 | 306 | |||
XM_008767125.1 | XP_008765347.1 | 2 | 307 | |||
XM_008767126.1 | XP_008765348.1 | 2 | 305 | |||
XM_008767127.1 | XP_008765349.1 | 2 | 306 |