Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TJP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170414.2 | NP_001163885.1 | 8 | 1248 | ||
NM_001170415.1 | NP_001163886.1 | 7 | 1248 | |||
NM_001170416.1 | NP_001163887.1 | 7 | 1178 | |||
NM_004817.3 | NP_004808.2 | 7 | 1403 | |||
NM_201629.3 | NP_963923.1 | 7 | 1403 | |||
XM_005252314.2 | XP_005252371.1 | 7 | 1456 | |||
XM_011519204.1 | XP_011517506.1 | 7 | 1146 | |||
XM_011519206.2 | XP_011517508.1 | 9 | 1748 | |||
XM_011519207.2 | XP_011517509.1 | 8 | 1244 | |||
XM_011519208.2 | XP_011517510.1 | 8 | 1204 | |||
XM_011519209.2 | XP_011517511.1 | 6 | 1033 | |||
XM_017015326.1 | XP_016870815.1 | 8 | 1301 | |||
XM_017015327.1 | XP_016870816.1 | 7 | 1403 | |||
XM_017015328.1 | XP_016870817.1 | 7 | 1456 | |||
GenBank mRNA | AF489824.1 | - | 7 | 1164 | ||
AK295016.1 | - | 8 | 1204 | |||
AK295034.1 | - | 7 | 1178 | |||
AK302483.1 | - | 7 | 1104 | |||
AK304447.1 | - | 9 | 1330 | |||
BC027592.1 | - | 7 | 1294 | 4839233 | ||
L27476.1 | - | 7 | 1164 |