Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 7 | 882 | ||
NM_001281502.1 | NP_001268431.1 | 7 | 951 | |||
NM_004265.3 | NP_004256.1 | 7 | 1223 | |||
GenBank mRNA | AF084559.1 | - | 7 | 1015 | ||
AF108658.1 | - | 7 | 808 | |||
AF126799.1 | - | 7 | 878 | |||
AK027459.1 | - | 1 | 18 | |||
AK027513.1 | - | 1 | 37 | |||
AK027577.1 | - | 1 | 62 | |||
AK074925.1 | - | 5 | 430 | |||
AK074939.1 | - | 7 | 955 | |||
AK074991.1 | - | 7 | 691 | |||
AK123240.1 | - | 2 | 852 | |||
AK290291.1 | - | 7 | 829 | |||
AK299762.1 | - | 7 | 951 | |||
AL050118.1 | - | 5 | 467 | |||
BC009011.1 | - | 7 | 957 | 4181037 |