Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004818.2 | NP_004809.2 | 10 | 1155 | ||
GenBank mRNA | AF026402.1 | - | 10 | 1103 | ||
AK128428.1 | - | 10 | 1084 | |||
AK292921.1 | - | 10 | 1144 | |||
AK294877.1 | - | 7 | 1019 | |||
AK312379.1 | - | 10 | 1124 | |||
BC002366.2 | - | 10 | 1114 | 2820898 |