Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100360856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244279.2 | XP_006244341.1 | 1 | 47 | ||
XM_008766796.1 | XP_008765018.1 | 29 |
Gene Symbol: LOC301748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770438.1 | XP_008768660.1 | 2 | 649 |
Gene Symbol: LOC302192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253275.2 | XP_006253337.1 | 29 |
Gene Symbol: LOC367381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079433.1 | - | 1 | 27 |
Gene Symbol: LOC681180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253262.2 | XP_006253324.1 | 1 | 29 |
Gene Symbol: LOC689268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252529.2 | XP_006252591.1 | 1 | 107 |
Gene Symbol: LOC689479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750610.3 | XP_003750658.2 | 1 | 373 | ||
XM_008766736.1 | XP_008764958.1 | 1 | 374 |
Gene Symbol: LOC691414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766795.1 | XP_008765017.1 | 1 | 101 |