Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244357.2 | XP_006244419.1 | 16 | 1510 | ||
XM_006244358.2 | XP_006244420.1 | 15 | 1482 | |||
XM_006244359.2 | XP_006244421.1 | 13 | 1324 | |||
XM_006244360.2 | XP_006244422.1 | 15 | 1305 | |||
XM_006244362.2 | XP_006244424.1 | 12 | 1119 | |||
XM_576696.6 | XP_576696.4 | 14 | 1277 | |||
GenBank mRNA | FQ212208.1 | - | 14 | 1943 |