Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004821.2 | NP_004812.1 | 1 | 660 | ||
XM_005268531.1 | XP_005268588.1 | 1 | 605 | |||
GenBank mRNA | AF061756.1 | - | 1 | 660 | ||
BC021190.2 | - | 1 | 629 | 3162118 |