Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004822.2 | NP_004813.2 | 6 | 1561 | ||
XM_006721595.3 | XP_006721658.1 | 6 | 1661 | |||
GenBank mRNA | GU727649.1 | - | 5 | 1461 | ||
U75586.1 | - | 6 | 1563 |