Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013135.2 | NP_001013153.2 | 4 | 492 | ||
XM_006233681.2 | XP_006233743.1 | 3 | 402 | |||
XM_006233682.2 | XP_006233744.1 | 3 | 321 | |||
XM_006233683.2 | XP_006233745.1 | 4 | 381 | |||
XM_006233684.2 | XP_006233746.1 | 2 | 234 | |||
XM_006233685.2 | XP_006233747.1 | 2 | 258 | |||
XM_006233686.2 | XP_006233748.1 | 4 | 471 | |||
GenBank mRNA | BC168170.1 | - | 2 | 264 | 9025284 |
Gene Symbol: LOC100363321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252792.1 | XP_006252854.1 | 1 | 257 |
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 326 |