Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004823.1 | NP_004814.1 | 3 | 1699 | ||
XM_011527526.1 | XP_011525828.1 | 4 | 2261 | |||
GenBank mRNA | AF117708.1 | - | 3 | 1699 | ||
AF134149.1 | - | 3 | 1708 | |||
AK022344.1 | - | 3 | 1428 | |||
BC004367.1 | - | 3 | 1629 | 3639657 |