Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024366.1 | NP_001019537.1 | 8 | 1067 | ||
XM_008773430.1 | XP_008771652.1 | 8 | 1588 | |||
GenBank mRNA | BC079325.1 | - | 8 | 1067 | 7135460 |