Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037216.1 | NP_001032293.1 | 6 | 695 | ||
XM_006257254.2 | XP_006257316.1 | 6 | 728 | |||
XM_008773431.1 | XP_008771653.1 | 6 | 1266 | |||
GenBank mRNA | BC100151.1 | - | 6 | 695 | 7456178 |