Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mum1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109321.1 | NP_001102791.1 | 1 | 2045 | ||
XM_006257301.2 | XP_006257363.1 | 5 | 2707 | |||
XM_006257302.2 | XP_006257364.1 | 6 | 2657 | |||
XM_006257303.2 | XP_006257365.1 | 5 | 2987 | |||
XM_006257304.2 | XP_006257366.1 | 4 | 2930 | |||
XM_006257305.2 | XP_006257367.1 | 5 | 2603 | |||
GenBank mRNA | BC168654.1 | - | 3 | 2497 | 7105971 |