Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100912478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751684.3 | XP_003751732.1 | 2 | 240 | ||
XM_006253887.2 | XP_006253949.1 | 3 | 511 |
Gene Symbol: LOC103689964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253890.2 | XP_006253952.1 | 511 | |||
XM_577054.6 | XP_577054.2 | 240 |
Gene Symbol: LOC103694079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771606.1 | XP_008769828.1 | 74 |
Gene Symbol: Vma21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253301.2 | XP_006253363.1 | 240 | |||
XM_008771344.1 | XP_008769566.1 | 511 | ||||
GenBank mRNA | FQ211836.1 | - | 204 | |||
FQ215711.1 | - | 506 |