Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109324.1 | NP_001102794.1 | 9, 10 | 996 | ||
XM_006229603.2 | XP_006229665.1 | 8, 9 | 1153 | |||
XM_006229604.2 | XP_006229666.1 | 8, 9 | 3565 | |||
XM_006229605.2 | XP_006229667.1 | 9, 10 | 2878 | |||
GenBank mRNA | BC169081.1 | - | 8, 9 | 1060 | 5621009 |